siRNA Details
Gene Symbol: GTF2IRD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003795.2 | NP_001003795.1 | 13 | 1237 | ||
GenBank mRNA | AK095572.1 | - | 4 | 289 | ||
AY312850.1 | - | 13 | 1212 | |||
AY312851.1 | - | 12 | 1137 | |||
BC067859.1 | - | 13 | 1202 | 30340785 | ||
BC136849.1 | - | 13 | 1197 | 9020840 | ||
BX647624.1 | - | 7 | 710 |
Gene Symbol: GTF2IRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173537.3 | NP_775808.3 | 13 | 1237 | ||
GenBank mRNA | AK095257.1 | - | 10 | 796 | ||
AK225845.1 | - | 13 | 1194 | |||
AK291431.1 | - | 13 | 1206 | |||
AL834153.1 | - | 1 | 87 | |||
AY260739.1 | - | 13 | 1190 | |||
AY312853.1 | - | 13 | 1190 | |||
AY312854.1 | - | 12 | 1030 |
Gene Symbol: GTF2IRD2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY312852.1 | - | 10 | 784 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
