siRNA Details
Gene Symbol: GTF2IRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173537.3 | NP_775808.3 | 16 | 2548 | ||
GenBank mRNA | AK095257.1 | - | 13 | 2108 | ||
AK225845.1 | - | 16 | 2505 | |||
AK291431.1 | - | 16 | 2517 | |||
AL833932.1 | - | 1 | 679 | |||
AL834153.1 | - | 3 | 1214 | |||
AY260739.1 | - | 16 | 2501 | |||
AY312853.1 | - | 16 | 2501 | |||
AY312854.1 | - | 15 | 2341 |
Gene Symbol: GTF2IRD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003795.2 | NP_001003795.1 | 16 | 2548 | ||
GenBank mRNA | AK025076.1 | - | 1 | 820 | ||
AK095572.1 | - | 7 | 1600 | |||
AY312850.1 | - | 16 | 2523 | |||
AY312851.1 | - | 15 | 2448 | |||
BC048252.1 | - | 1 | 820 | |||
BC136849.1 | - | 16 | 2508 | 9020840 | ||
BX647624.1 | - | 10 | 2021 |
Gene Symbol: GTF2IRD2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY312852.1 | - | 13 | 2094 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
