siRNA Details
Gene Symbol: PDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161779.1 | NP_001155251.1 | 3 | 580 | ||
NM_001161780.1 | NP_001155252.1 | 3 | 454 | |||
NM_001161781.1 | NP_001155253.1 | 2 | 441 | |||
NM_018444.3 | NP_060914.2 | 2 | 498 | |||
XM_011517135.2 | XP_011515437.1 | 3 | 351 | |||
XM_011517136.1 | XP_011515438.1 | 3 | 458 | |||
XM_017013587.1 | XP_016869076.1 | 2 | 513 | |||
XM_017013588.1 | XP_016869077.1 | 3 | 421 | |||
GenBank mRNA | AF155661.1 | - | 3 | 468 | ||
AK057079.1 | - | 2 | 410 | |||
AK126862.1 | - | 1 | 875 | |||
AK302651.1 | - | 2 | 384 | |||
BC047619.1 | - | 2 | 419 | 4824168 | ||
BC064978.1 | - | 3 | 438 | |||
BC098343.1 | - | 1 | 252 | 40010016 | ||
DA240240.1 | - | 2 | 443 | |||
DA507397.1 | - | 3 | 454 | |||
DB084539.1 | - | 2 | 498 | |||
DC369809.1 | - | 2 | 455 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
