siRNA Details
Gene Symbol: NPSR1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_015356.2 | 3 | 303 | |||
NR_033664.1 | 2 | 232 | ||||
NR_033665.1 | 2 | 232 | ||||
GenBank mRNA | AY312364.1 | - | 3 | 306 | ||
AY312365.1 | - | LIT3317 AAA1 |
3 | 301 | ||
AY312366.1 | - | LIT3326 AAA1 |
3 | 197 | ||
AY312367.1 | - | LIT3327 AAA1 |
2 | 232 | ||
AY312368.1 | - | LIT3328 AAA1 |
2 | 232 | ||
AY312369.1 | - | LIT3332 AAA1 |
2 | 232 | ||
AY312370.1 | - | LIT3333 AAA1 |
2 | 117 | ||
AY312371.1 | - | LIT3334 AAA1 |
2 | 232 | ||
AY312372.1 | - | LIT3335 AAA1 |
2 | 232 | ||
AY312373.1 | - | LIT3331 AAA1 |
3 | 197 | ||
AY321515.1 | - | 2 | 113 | |||
AY321516.1 | - | 2 | 113 | |||
BC031961.1 | - | 3 | 284 | |||
BC130477.1 | - | 2 | 117 | |||
U85653.1 | - | 2 | 195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
