siRNA Details
Gene Symbol: NEK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126330.1 | - | 9 | 2737 |
Gene Symbol: FKSG49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336878.1 | - | 1 | 372 | ||
AF336883.1 | - | 1 | 381 | |||
AF336884.1 | - | 1 | 441 | |||
AF336886.1 | - | 1 | 369 | |||
AF336887.1 | - | 1 | 426 | |||
AF338190.1 | - | 1 | 414 | |||
AF338194.1 | - | 1 | 435 | |||
AF338195.1 | - | 1 | 432 | |||
AF338196.1 | - | 1 | 444 | |||
AF338199.1 | - | 1 | 399 | |||
AF469044.1 | - | 1 | 381 |
Gene Symbol: LOC402469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF336877.1 | - | 1 | 444 |
Gene Symbol: SQSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK094484.1 | - | 1 | 106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
