siRNA Details
Gene Symbol: RPL41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035267.1 | NP_001030344.1 | 3 | 493 | ||
NM_021104.1 | NP_066927.1 | 4 | 378 | |||
GenBank mRNA | AF026844.1 | - | 1 | 339 | ||
BC014383.1 | - | 1 | 340 | |||
BC015984.1 | - | 1 | 339 | |||
BC017820.1 | - | 3 | 448 | |||
BC032611.1 | - | 1 | 330 | |||
BC062462.1 | - | 1 | 321 | |||
BC064907.1 | - | 1 | 321 | |||
BC066328.1 | - | 3 | 405 | |||
BC070252.1 | - | 1 | 336 | |||
BC070253.1 | - | 1 | 335 | |||
BC070254.1 | - | 1 | 336 | |||
BC070255.1 | - | 1 | 335 | |||
BC070483.1 | - | 1 | 302 | |||
BC082759.1 | - | 3 | 433 | |||
BC105279.1 | - | 1 | 339 | |||
BC105286.1 | - | 1 | 340 | |||
BC105296.1 | - | 1 | 340 | |||
BC105794.1 | - | 1 | 339 | |||
BC105795.1 | - | 1 | 339 | |||
BC105799.1 | - | 1 | 340 | |||
BC142666.1 | - | 1 | 319 | |||
BC142726.1 | - | 1 | 319 | |||
Z12962.1 | - | 4 | 378 |
Gene Symbol: CTTNBP2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK023340.1 | - | 1746 |
Gene Symbol: RPL41P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | S64030.1 | - | 1 | 255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
