siRNA Details
Gene Symbol: SPAG11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081552.2 | NP_001075021.2 | 3 | 401 | ||
XM_005272391.4 | XP_005272448.2 | 3 | 374 | |||
XM_017013744.1 | XP_016869233.1 | 2 | 75 | |||
GenBank mRNA | AF170797.1 | - | 3 | 213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
