siRNA Details
Gene Symbol: ALDH6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278593.1 | NP_001265522.1 | 12 | 2627 | ||
NM_001278594.1 | NP_001265523.1 | 12 | 2829 | |||
NM_005589.3 | NP_005580.1 | 12 | 2666 | |||
XM_017021331.1 | XP_016876820.1 | 10 | 2517 |
Gene Symbol: AMY2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DQ786288.1 | - | 3, 4 | 359 |
Gene Symbol: ANGEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300753.1 | NP_001287682.1 | 9 | 2535 | ||
NM_001300755.1 | NP_001287684.1 | 9 | 2633 | |||
NM_001300757.1 | NP_001287686.1 | 8 | 2288 | |||
NM_001300758.1 | NP_001287687.1 | 8 | 2284 | |||
NM_144567.4 | NP_653168.2 | 9 | 2610 | |||
NR_125333.1 | 7 | 2157 | ||||
XM_005273344.1 | XP_005273401.1 | 9 | 2631 | |||
XM_005273345.1 | XP_005273402.1 | 9 | 2677 | |||
XM_005273346.2 | XP_005273403.1 | 9 | 2430 | |||
XM_005273347.3 | XP_005273404.1 | 8 | 2302 | |||
XM_017002774.1 | XP_016858263.1 | 9 | 2559 | |||
XM_017002775.1 | XP_016858264.1 | 10 | 2858 | |||
XM_017002776.1 | XP_016858265.1 | 10 | 2882 | |||
XM_017002777.1 | XP_016858266.1 | 10 | 2928 | |||
XM_017002778.1 | XP_016858267.1 | 10 | 2671 | |||
GenBank mRNA | AF510741.1 | - | 8 | 2239 | ||
AK022855.1 | - | 1 | 601 | |||
BC047469.1 | - | 8 | 2201 | 5274835 |
Gene Symbol: C18orf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055900.1 | - | 1 | 1418 |
Gene Symbol: COX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011524342.2 | XP_011522644.1 | 5 | 5572 | ||
XM_017024192.1 | XP_016879681.1 | 5 | 5576 | |||
XM_017024194.1 | XP_016879683.1 | 5 | 1725 | |||
XM_017024195.1 | XP_016879684.1 | 5 | 1729 |
Gene Symbol: CPT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001876.3 | NP_001867.2 | 19 | 4036 | ||
XM_005273762.2 | XP_005273819.1 | 19 | 4012 | |||
GenBank mRNA | AJ420378.1 | - | 1 | 1631 |
Gene Symbol: CROCCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090414.1 | - | 11 | 3801 |
Gene Symbol: DUSP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK226170.1 | - | 1, 2 | 251 |
Gene Symbol: ERAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537542.1 | - | 3 | 1060 |
Gene Symbol: FAM83G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123558.1 | - | 3 | 4419 |
Gene Symbol: FFAR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI469384.1 | - | 1 | 130 |
Gene Symbol: FKBP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143782.1 | NP_001137254.1 | 6 | 976 | ||
GenBank mRNA | AK094876.1 | - | 2 | 1192 | ||
AK301455.1 | - | 6 | 949 | |||
BC048292.1 | - | 4 | 1489 | 5207367 |
Gene Symbol: GRIK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC037954.1 | - | 6, 7 | 1446 |
Gene Symbol: HSD17B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127832.1 | - | 1 | 156 |
Gene Symbol: IMPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC031560.1 | - | 7 | 1260 |
Gene Symbol: INPP5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297434.1 | NP_001284363.1 | 18 | 2926 | ||
NM_005540.2 | NP_005531.2 | 24 | 3433 | |||
XM_006710621.3 | XP_006710684.2 | 20 | 3196 | |||
XM_011541392.2 | XP_011539694.1 | 18 | 2872 | |||
XM_011541393.2 | XP_011539695.1 | 18 | 2861 | |||
XM_017001207.1 | XP_016856696.1 | 21 | 3173 | |||
XM_017001208.1 | XP_016856697.1 | 21 | 3302 | |||
XM_017001209.1 | XP_016856698.1 | 18 | 2852 |
Gene Symbol: IPO5P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103741.1 | 1, 2 | 186 | |||
GenBank mRNA | AK023040.1 | - | 1, 2 | 165 | ||
DA185451.1 | - | 1, 2 | 171 |
Gene Symbol: LOC100505549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242804.1 | NP_001229733.1 | 2 | 582 | ||
GenBank mRNA | BC040476.1 | - | 2 | 535 |
Gene Symbol: LOC101929352 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125507.1 | - | 2 | 566 |
Gene Symbol: LOC283278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833334.1 | - | 1 | 2427 | ||
AL833346.1 | - | 1 | 2427 |
Gene Symbol: LRWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209379.1 | - | 7 | 3283 |
Gene Symbol: MLX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209235.1 | - | 953 |
Gene Symbol: MROH7-TTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037639.2 | 3, 4 | 1684 | |||
GenBank mRNA | CR749830.1 | - | 3, 4 | 1636 |
Gene Symbol: MRPS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK058101.1 | - | 1 | 473 |
Gene Symbol: NME6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BQ020663.1 | - | 1 | 282 |
Gene Symbol: PARP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017554.2 | NP_060024.2 | 17 | 6332 | ||
GenBank mRNA | AK022542.1 | - | 5 | 2134 | ||
AK027260.1 | - | 1 | 1137 | |||
AY134858.1 | - | 12 | 5505 | |||
BX648758.1 | - | 13 | 6915 |
Gene Symbol: PATJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011540470.1 | XP_011538772.1 | 20, 21 | 2957 |
Gene Symbol: PDE12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322177.1 | NP_001309106.1 | 2 | 2487 | ||
NM_177966.6 | NP_808881.3 | 3 | 2314 | |||
GenBank mRNA | AK001526.1 | - | 1 | 583 | ||
AK300374.1 | - | 3 | 2287 | |||
AL831824.1 | - | 3 | 1818 | |||
BX647169.1 | - | 3 | 2280 |
Gene Symbol: PLEKHA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC032761.1 | - | 1 | 133 |
Gene Symbol: PLEKHM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123204.1 | - | 6, 7 | 1428 |
Gene Symbol: PPP2R3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC037797.1 | - | 1 | 1399 |
Gene Symbol: PRKCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002737.2 | NP_002728.1 | 17 | 6766 | ||
XM_017024837.1 | XP_016880326.1 | 16 | 6597 | |||
XM_017024838.1 | XP_016880327.1 | 17 | 6693 | |||
XM_017024839.1 | XP_016880328.1 | 16 | 7632 | |||
XM_017024840.1 | XP_016880329.1 | 17 | 6675 | |||
GenBank mRNA | AK055431.1 | - | 1 | 419 | ||
AK125425.1 | - | 1 | 1109 | |||
BX648954.1 | - | 1 | 3733 |
Gene Symbol: PRKCSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC015154.1 | - | 7 | 1058 |
Gene Symbol: RAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK122742.1 | - | 1 | 270 |
Gene Symbol: RDH13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011526408.2 | XP_011524710.1 | 7 | 2584 | ||
GenBank mRNA | AK096798.1 | - | 1 | 1990 |
Gene Symbol: RUFY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017987.4 | NP_060457.4 | 18 | 3349 | ||
XM_005269953.4 | XP_005270010.1 | 17 | 3247 | |||
XM_005269955.4 | XP_005270012.1 | 17 | 2928 | |||
XM_005269956.4 | XP_005270013.1 | 17 | 3181 | |||
XM_005269957.4 | XP_005270014.1 | 17 | 2917 | |||
XM_006717911.3 | XP_006717974.1 | 18 | 3020 | |||
XM_011539942.2 | XP_011538244.1 | 18 | 3761 | |||
GenBank mRNA | AK055646.1 | - | 1 | 1137 | ||
AK091629.1 | - | 2 | 1958 | |||
AK127019.1 | - | 18 | 3350 |
Gene Symbol: SCML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006089.2 | NP_006080.1 | 15 | 3542 | ||
XM_006724458.1 | XP_006724521.1 | 14 | 3354 | |||
XM_017029221.1 | XP_016884710.1 | 14 | 3424 | |||
GenBank mRNA | AK022824.1 | - | 3 | 1790 | ||
BC064617.1 | - | 15 | 3499 | 5575894 |
Gene Symbol: SP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080391.1 | NP_001073860.1 | 29 | 4370 | ||
GenBank mRNA | AK090565.1 | - | 1 | 1576 | ||
AK160379.1 | - | 5 | 3760 |
Gene Symbol: STOM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270526.1 | NP_001257455.1 | 7 | 1357 | ||
NM_004099.5 | NP_004090.4 | 7 | 1448 | |||
NM_198194.2 | NP_937837.1 | 3 | 953 | |||
NR_073037.1 | 8 | 1578 | ||||
GenBank mRNA | AK304518.1 | - | 6 | 1091 | ||
BC010703.1 | - | 7 | 1369 | 3873494 | ||
BM925356.1 | - | 1 | 530 | |||
M81635.1 | - | 7 | 1353 |
Gene Symbol: SVIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095023.1 | - | 1 | 1815 |
Gene Symbol: THBS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA755868.1 | - | 1, 2 | 194 |
Gene Symbol: TMX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021156.3 | NP_066979.2 | 8 | 3792 | ||
GenBank mRNA | AB032988.1 | - | 1 | 2876 | ||
BC044777.1 | - | 8 | 3791 |
Gene Symbol: TSPAN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126702.1 | - | 1 | 428 |
Gene Symbol: UTY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047603.1 | 2, 3 | 1231 | |||
GenBank mRNA | EF653186.1 | - | 2, 3 | 226 |
Gene Symbol: ZBTB25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304507.1 | NP_001291436.1 | 3, 4 | 1211 | ||
XM_011537143.2 | XP_011535445.1 | 1 | 1055 | |||
XM_017021632.1 | XP_016877121.1 | 3, 4 | 493 | |||
GenBank mRNA | AK098181.1 | - | 3, 4 | 582 |
Gene Symbol: ZNF274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095162.1 | - | 1 | 880 | ||
BX641169.1 | - | 1 | 2650 |
Gene Symbol: ZNF738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027130.1 | 5 | 9879 |
Gene Symbol: ZNF765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040185.1 | NP_001035275.1 | 4 | 3890 | ||
GenBank mRNA | BX647981.1 | - | 4 | 3887 |
Gene Symbol: ZNF783 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195220.1 | NP_001182149.1 | 6 | 2461 | ||
XM_011515701.2 | XP_011514003.1 | 6 | 2389 | |||
GenBank mRNA | AK022976.1 | - | 1 | 483 | ||
AL359620.1 | - | 1 | 987 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
