siRNA Details
Gene Symbol: CCNDBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012142.4 | NP_036274.3 | 11 | 2919 | ||
NR_027513.2 | 9 | 2742 | ||||
NR_027514.2 | 10 | 2822 | ||||
NR_045998.1 | 10 | 2839 | ||||
NR_045999.1 | 8 | 2682 |
Gene Symbol: RAB40B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005256335.3 | XP_005256392.1 | 3 | 1299 | ||
GenBank mRNA | AK124376.1 | - | 1 | 644 |
Gene Symbol: SLC35B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648022.1 | - | 2 | 3875 |
Gene Symbol: SMYD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003163.1 | XP_016858652.1 | 1 | 95 |
Gene Symbol: ZNF250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648173.1 | - | 3 | 2550 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
