siRNA Details
Gene Symbol: TNS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032865.5 | NP_116254.4 | 13 | 3093 | ||
XM_005257744.1 | XP_005257801.1 | 13 | 3169 | |||
XM_017025236.1 | XP_016880725.1 | 12 | 3094 | |||
GenBank mRNA | AF370421.1 | - | 13 | 3087 | ||
AF417488.1 | - | 13 | 3054 | |||
AK027856.1 | - | 11 | 2442 | |||
AW937802.1 | - | 1 | 310 | |||
BC013373.2 | - | 4 | 1205 | |||
BC018706.1 | - | 12 | 2539 | 5013235 |
Gene Symbol: C21orf59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FJ649644.1 | - | 1 | 91 |
Gene Symbol: CDC14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF064105.2 | - | 14 | 2513 |
Gene Symbol: LOC285389 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136181.1 | 3 | 2145 | |||
GenBank mRNA | BC038368.1 | - | 3 | 2145 |
Gene Symbol: MTSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011517404.2 | XP_011515706.1 | 1 | 11762 | ||
XM_017014101.1 | XP_016869590.1 | 1 | 255 |
Gene Symbol: NASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092829.1 | - | 1 | 981 |
Gene Symbol: PKD1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832135.1 | - | 1 | 2458 |
Gene Symbol: SNRNP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104103.1 | 2 | 166 |
Gene Symbol: SP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126529.1 | - | 1 | 247 |
Gene Symbol: SRSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078166.1 | NP_001071634.1 | 3 | 3606 | ||
NM_006924.4 | NP_008855.1 | 4 | 3406 | |||
NR_034041.1 | 4 | 2685 |
Gene Symbol: TIMM10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012192.3 | NP_036324.1 | 3 | 2323 |
Gene Symbol: ZNF503-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038226.1 | 2 | 323 | |||
GenBank mRNA | CN360505.1 | - | 2 | 209 |
Gene Symbol: ZNF836 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011526558.2 | XP_011524860.1 | 5 | 3443 | ||
XM_011526559.2 | XP_011524861.1 | 4 | 3488 | |||
XM_017026389.1 | XP_016881878.1 | 5 | 3376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
