siRNA Details
Gene Symbol: TNXB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019105.6 | NP_061978.6 | 34 | 11518 | ||
NM_032470.3 | NP_115859.2 | 3 | 1518 | |||
GenBank mRNA | AB209012.1 | - | 5 | 1244 | ||
BC033740.1 | - | 5 | 1095 | |||
BC071883.1 | - | 4 | 1149 | |||
BC125114.1 | - | 3 | 1420 | |||
BC125115.1 | - | 3 | 1420 | |||
BC130037.1 | - | 3 | 1420 | |||
EU219621.1 | - | 3 | 481 | |||
M25813.1 | - | 5 | 1050 | |||
U24488.1 | - | 3 | 1518 |
Gene Symbol: TNXA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001284.2 | 3 | 1267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
