siRNA Details
Gene Symbol: IGKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC067092.1 | - | 2 | 208 | ||
BC073764.1 | - | 2 | 199 | |||
BC073791.1 | - | 2 | 204 | |||
BC095489.1 | - | 2 | 208 |
Gene Symbol: IGKV@ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF103383.1 | - | 2 | 111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
