siRNA Details
Gene Symbol: TBXA2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001060.5 | NP_001051.1 | 3 | 2062 | ||
NM_201636.2 | NP_963998.2 | 4 | 1402 | |||
GenBank mRNA | BC121148.1 | - | 2 | 1321 | 40121265 | |
U11271.1 | - | 4 | 1226 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
