siRNA Details
Gene Symbol: TBXA2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001060.5 | NP_001051.1 | 2 | 1121 | ||
NM_201636.2 | NP_963998.2 | 2 | 1121 | |||
XM_011528214.2 | XP_011526516.1 | 3 | 3155 | |||
GenBank mRNA | AY429110.1 | - | 1 | 734 | ||
BC074749.2 | - | 1 | 773 | 30915459 | ||
BC074750.2 | - | 1 | 773 | 30915209 | ||
U11271.1 | - | 2 | 946 | |||
U27325.1 | - | 2 | 857 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
