siRNA Details
Gene Symbol: TBXA2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001060.5 | NP_001051.1 | 1 | 199 | ||
NM_201636.2 | NP_963998.2 | 1 | 199 | |||
XM_011528214.2 | XP_011526516.1 | 1 | 2115 | |||
GenBank mRNA | BC028027.1 | - | 1 | 199 | ||
U11271.1 | - | 1 | 25 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
