siRNA Details
Gene Symbol: SHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010846.2 | NP_001010846.1 | 6 | 1807 | ||
NR_135169.1 | 6 | 1802 | ||||
GenBank mRNA | AK074067.1 | - | 7 | 1226 | ||
AK126006.1 | - | 4 | 1072 | |||
CR936667.1 | - | 6 | 1250 | |||
CR936736.1 | - | 3 | 2482 |
Gene Symbol: BLOC1S1-RDH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096656.1 | - | 4 | 1359 |
Gene Symbol: CECR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290046.1 | NP_001276975.1 | 19 | 8811 | ||
NM_001290047.1 | NP_001276976.1 | 19 | 9283 | |||
XM_006724079.3 | XP_006724142.1 | 7 | 7726 | |||
XM_011546128.2 | XP_011544430.1 | 18 | 8887 | |||
XM_011546129.2 | XP_011544431.1 | 19 | 8887 | |||
XM_011546132.2 | XP_011544434.1 | 19 | 8806 | |||
XM_017028785.1 | XP_016884274.1 | 19 | 8968 | |||
GenBank mRNA | AK055696.1 | - | 1 | 758 |
Gene Symbol: CLSPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190481.1 | NP_001177410.1 | 24 | 5867 | ||
NM_022111.3 | NP_071394.2 | 25 | 6059 |
Gene Symbol: DET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090766.1 | - | 2 | 1337 |
Gene Symbol: LINC00926 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024433.1 | 1 | 88 | |||
GenBank mRNA | AK097083.1 | - | 1 | 88 | ||
DB117396.1 | - | 1 | 88 |
Gene Symbol: LOC105372547 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC041905.1 | - | 1 | 1796 |
Gene Symbol: OTULIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138348.4 | NP_612357.4 | 7 | 5501 | ||
XM_011514154.2 | XP_011512456.1 | 6 | 9111 | |||
XM_017010015.1 | XP_016865504.1 | 6 | 9304 | |||
GenBank mRNA | AK095615.1 | - | 1 | 1388 | ||
AK123480.1 | - | 1 | 2483 |
Gene Symbol: PCBP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056822.1 | - | 5 | 899 |
Gene Symbol: PDLIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK055582.1 | - | 1 | 197 | ||
AK074031.1 | - | 2 | 3302 |
Gene Symbol: SENP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308045.1 | NP_001294974.1 | 9 | 5218 | ||
NM_152699.4 | NP_689912.2 | 10 | 5356 | |||
XM_005269310.2 | XP_005269367.1 | 9 | 5272 | |||
XM_005269312.2 | XP_005269369.1 | 8 | 5134 | |||
XM_011512544.2 | XP_011510846.1 | 11 | 5401 | |||
XM_017005870.1 | XP_016861359.1 | 10 | 5218 | |||
GenBank mRNA | AL049229.2 | - | 1 | 749 | ||
BC013799.1 | - | 1 | 814 | |||
BC036626.1 | - | 1 | 389 |
Gene Symbol: SUGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011534891.2 | XP_011533193.1 | 14 | 10752 | ||
XM_011534893.2 | XP_011533195.1 | 13 | 10470 | |||
XM_017020365.1 | XP_016875854.1 | 14 | 10642 | |||
XM_017020366.1 | XP_016875855.1 | 14 | 10517 | |||
XM_017020367.1 | XP_016875856.1 | 14 | 10551 | |||
XM_017020368.1 | XP_016875857.1 | 14 | 10563 |
Gene Symbol: VKORC1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011515830.2 | XP_011514132.1 | 1 | 1387 |
Gene Symbol: WEE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX641032.1 | - | 4 | 2713 |
Gene Symbol: ZBTB8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040441.2 | NP_001035531.2 | 5 | 4131 | ||
NM_001291496.1 | NP_001278425.1 | 5 | 3950 | |||
NR_111980.1 | 4 | 3126 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
