siRNA Details
Gene Symbol: BRI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017011936.1 | XP_016867425.1 | 3 | 3120 |
Gene Symbol: AHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001621.4 | NP_001612.1 | 11 | 4809 | ||
GenBank mRNA | AA329775.1 | - | 1 | 236 | ||
L19872.1 | - | 11 | 4578 |
Gene Symbol: AQP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001652.3 | NP_001643.2 | 4 | 1833 | ||
GenBank mRNA | AL137716.1 | - | 7 | 4308 |
Gene Symbol: BBS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB208905.1 | - | 8 | 3068 | ||
AK127290.1 | - | 1 | 362 |
Gene Symbol: BET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133908.1 | 7 | 2375 | |||
NR_133909.1 | 5 | 2252 | ||||
GenBank mRNA | CR749267.1 | - | 7 | 2327 |
Gene Symbol: CAPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011544286.2 | XP_011542588.1 | 12 | 1958 | ||
GenBank mRNA | AK124751.1 | - | 1 | 324 |
Gene Symbol: COX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124809.1 | - | 1 | 2402 |
Gene Symbol: FAM3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282311.1 | NP_001269240.1 | 3 | 636 | ||
XM_005274714.3 | XP_005274771.1 | 4 | 486 | |||
XM_005274716.3 | XP_005274773.1 | 4 | 486 | |||
XM_006724831.2 | XP_006724894.1 | 4 | 486 | |||
XM_006724833.2 | XP_006724896.1 | 4 | 486 | |||
XM_006724834.2 | XP_006724897.1 | 3 | 544 | |||
XM_017029702.1 | XP_016885191.1 | 2 | 60 | |||
GenBank mRNA | AL539139.3 | - | 3 | 633 | ||
BC002934.1 | - | 3 | 540 |
Gene Symbol: GS1-124K5.11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK024371.1 | - | 4555 |
Gene Symbol: GSDMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057588.1 | - | 1 | 77 |
Gene Symbol: HYKK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013619.3 | NP_001013641.2 | 5 | 3366 |
Gene Symbol: KLF3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026804.1 | 6 | 926 | |||
GenBank mRNA | AK023259.1 | - | 6 | 926 |
Gene Symbol: LOC730668 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027240.1 | 1 | 2167 | |||
GenBank mRNA | AK128491.1 | - | 1 | 2179 |
Gene Symbol: LRRC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011523204.1 | XP_011521506.1 | 3 | 336 | ||
GenBank mRNA | AK001866.1 | - | 3 | 368 |
Gene Symbol: MAGI2-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038343.2 | 4 | 4157 | |||
GenBank mRNA | BX641113.1 | - | 1 | 3050 | ||
CR749529.1 | - | 2 | 4210 |
Gene Symbol: MTHFD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011535738.2 | XP_011534040.1 | 21 | 2234 |
Gene Symbol: MTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010891.4 | NP_001010891.4 | 8 | 3454 | ||
NM_001167741.1 | NP_001161213.1 | 8 | 3473 | |||
XM_005248495.3 | XP_005248552.1 | 9 | 3545 | |||
XM_017009440.1 | XP_016864929.1 | 7 | 3378 | |||
GenBank mRNA | BC035085.1 | - | 1 | 1708 | ||
BC150295.1 | - | 8 | 3447 | |||
BX538064.1 | - | 8 | 3456 | |||
BX647596.1 | - | 6 | 1780 |
Gene Symbol: NHLRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198514.3 | NP_940916.2 | 11 | 4170 | ||
GenBank mRNA | AL832090.1 | - | 2 | 2141 | ||
BX538287.1 | - | 7 | 2964 | |||
BX647641.1 | - | 9 | 3541 |
Gene Symbol: NPC1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013389.2 | NP_037521.2 | 15 | 3256 | ||
GenBank mRNA | AF192522.1 | - | 15 | 3256 |
Gene Symbol: RAD51-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000890.1 | - | 1 | 864 |
Gene Symbol: RHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127691.2 | NP_001121163.1 | 7 | 1432 | ||
NM_001282867.1 | NP_001269796.1 | 10 | 1716 | |||
NM_001282868.1 | NP_001269797.1 | 8 | 1512 | |||
NM_001282869.1 | NP_001269798.1 | 8 | 1566 | |||
NM_001282870.1 | NP_001269799.1 | 9 | 1646 | |||
NM_001282871.1 | NP_001269800.1 | 9 | 1736 | |||
NM_001282872.1 | NP_001269801.1 | 10 | 1816 | |||
NM_016124.4 | NP_057208.2 | 10 | 1720 | |||
GenBank mRNA | X63097.1 | - | 10 | 1678 |
Gene Symbol: SLC35F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005027.1 | XP_016860516.1 | 17 | 1907 |
Gene Symbol: WDR37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128194.1 | - | 1 | 1963 |
Gene Symbol: ZFPM2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125796.1 | 3 | 238 | |||
NR_125797.1 | 3 | 249 | ||||
GenBank mRNA | BF244158.1 | - | 3 | 214 | ||
BF977562.1 | - | 125 | ||||
BU959299.1 | - | 213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
