siRNA Details
Gene Symbol: CFLAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127183.2 | NP_001120655.1 | 10 | 3998 | ||
NM_001202515.1 | NP_001189444.1 | 6 | 3064 | |||
NM_001202516.1 | NP_001189445.1 | 8 | 3865 | |||
NM_001202517.1 | NP_001189446.1 | 9 | 3552 | |||
NM_001202518.1 | NP_001189447.1 | 8 | 4127 | |||
NM_001202519.1 | NP_001189448.1 | 9 | 4391 | |||
NM_003879.5 | NP_003870.4 | 10 | 3970 | |||
XM_017005189.1 | XP_016860678.1 | 11 | 4865 | |||
XM_017005190.1 | XP_016860679.1 | 10 | 3834 | |||
XM_017005191.1 | XP_016860680.1 | 10 | 3913 | |||
XM_017005192.1 | XP_016860681.1 | 10 | 4800 | |||
XM_017005193.1 | XP_016860682.1 | 10 | 6817 | |||
XM_017005194.1 | XP_016860683.1 | 10 | 4673 | |||
XM_017005195.1 | XP_016860684.1 | 11 | 4011 | |||
XM_017005196.1 | XP_016860685.1 | 8 | 3636 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
