siRNA Details
Gene Symbol: LGALS9C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040078.2 | NP_001035167.2 | 9 | 797 | ||
XM_005256769.1 | XP_005256826.1 | 9 | 794 | |||
XM_005256770.1 | XP_005256827.1 | 8 | 698 | |||
XM_011523993.1 | XP_011522295.1 | 8 | 701 | |||
XM_011523994.1 | XP_011522296.1 | 7 | 665 | |||
XM_011523995.1 | XP_011522297.1 | 9 | 797 | |||
XM_011523996.1 | XP_011522298.1 | 8 | 701 | |||
XM_011523997.1 | XP_011522299.1 | 7 | 435 | |||
GenBank mRNA | AK315818.1 | - | 9 | 784 | ||
BC073889.1 | - | 9 | 766 | 5222492 | ||
BM769737.1 | - | 5 | 221 |
Gene Symbol: LGALS9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042685.1 | NP_001036150.1 | 9 | 789 | ||
GenBank mRNA | AK290263.1 | - | 9 | 793 | ||
AK296659.1 | - | 8 | 706 | |||
BC105943.1 | - | 9 | 769 | 40031793 | ||
BM769345.1 | - | 5 | 221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
