siRNA Details
Gene Symbol: ANKRD20A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012419.2 | NP_001012419.1 | 15 | 2377 | ||
XM_005272525.3 | XP_005272582.1 | 15 | 2630 | |||
XM_011543860.2 | XP_011542162.1 | 15 | 2634 | |||
GenBank mRNA | BC140809.1 | - | 15 | 2374 | 9021677 | |
BC171793.1 | - | 15 | 2374 | 9054447 |
Gene Symbol: ANKRD20A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032250.3 | NP_115626.2 | 15 | 2411 | ||
XM_005272532.4 | XP_005272589.1 | 15 | 2345 | |||
GenBank mRNA | AL136793.1 | - | 15 | 2412 |
Gene Symbol: ANKRD20A12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046228.1 | 1 | 523 | |||
GenBank mRNA | AK090412.1 | - | 1 | 1524 | ||
BC022380.1 | - | 1 | 523 |
Gene Symbol: ANKRD20A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012421.1 | NP_001012421.1 | 15 | 2411 | ||
XM_011517877.2 | XP_011516179.1 | 15 | 2411 | |||
XM_011517878.2 | XP_011516180.1 | 15 | 2411 |
Gene Symbol: ANKRD20A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098805.1 | NP_001092275.1 | 15 | 2411 |
Gene Symbol: ANKRD20A5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040113.1 | 16 | 2597 |
Gene Symbol: ANKRD20A8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003366.2 | 13 | 2116 | |||
GenBank mRNA | CR936649.1 | - | 2 | 1431 |
Gene Symbol: ANKRD20A9P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027995.1 | 1 | 36473 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
