siRNA Details
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310136.1 | NP_001297065.1 | 5 | 629 | ||
XM_017023622.1 | XP_016879111.1 | 6 | 753 | |||
XM_017023623.1 | XP_016879112.1 | 6 | 753 | |||
XM_017023624.1 | XP_016879113.1 | 6 | 753 | |||
XM_017023625.1 | XP_016879114.1 | 6 | 733 | |||
XM_017023626.1 | XP_016879115.1 | 7 | 1372 | |||
XM_017023627.1 | XP_016879116.1 | 6 | 753 |
Gene Symbol: LOC613037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321892.1 | NP_001308821.1 | 7 | 768 | ||
XM_017023514.1 | XP_016879003.1 | 7 | 1057 | |||
XM_017023515.1 | XP_016879004.1 | 8 | 1076 | |||
XM_017023516.1 | XP_016879005.1 | 7 | 1103 | |||
XM_017023517.1 | XP_016879006.1 | 7 | 834 | |||
XM_017023518.1 | XP_016879007.1 | 7 | 922 | |||
XM_017023519.1 | XP_016879008.1 | 7 | 979 | |||
XM_017023520.1 | XP_016879009.1 | 2 | 2043 | |||
XM_017023521.1 | XP_016879010.1 | 3 | 298 |
Gene Symbol: NPIPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310137.1 | NP_001297066.1 | 5 | 575 | ||
GenBank mRNA | KF175195.1 | - | 5 | 479 | ||
KF175204.1 | - | 5 | 518 |
Gene Symbol: NPIPB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306094.1 | NP_001293023.1 | 5 | 597 | ||
XM_011523496.2 | XP_011521798.1 | 6 | 653 | |||
XM_011523498.2 | XP_011521800.1 | 6 | 687 | |||
XM_011523501.2 | XP_011521803.1 | 2 | 479 | |||
XM_017023248.1 | XP_016878737.1 | 6 | 1090 | |||
XM_017023249.1 | XP_016878738.1 | 5 | 907 |
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128772.1 | - | 1 | 1771 | ||
KF175196.1 | - | 5 | 479 | |||
KF175205.1 | - | 5 | 575 | |||
KF175206.1 | - | 5 | 575 | |||
KF175218.1 | - | 5 | 480 |
Gene Symbol: NPIPB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK296301.1 | - | 135 |
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282524.1 | NP_001269453.1 | 5 | 667 | ||
XM_005255741.3 | XP_005255798.1 | 6 | 1479 | |||
XM_005255742.4 | XP_005255799.1 | 6 | 1225 | |||
XM_005255743.3 | XP_005255800.1 | 2 | 2058 | |||
XM_011545929.2 | XP_011544231.1 | 7 | 1299 | |||
XM_011545931.2 | XP_011544233.1 | 6 | 932 | |||
XM_017023628.1 | XP_016879117.1 | 6 | 682 | |||
XM_017023629.1 | XP_016879118.1 | 6 | 1225 | |||
XM_017023630.1 | XP_016879119.1 | 7 | 952 | |||
XM_017023631.1 | XP_016879120.1 | 5 | 1296 | |||
XM_017023632.1 | XP_016879121.1 | 6 | 682 | |||
GenBank mRNA | BC144439.1 | - | 5 | 628 | ||
BC157836.1 | - | 5 | 1317 | 9057055 | ||
KF175222.1 | - | 5 | 629 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287250.1 | NP_001274179.1 | 5 | 667 | ||
NM_001287251.1 | NP_001274180.1 | 6 | 753 | |||
XM_003118719.7 | XP_003118767.2 | 6 | 1228 | |||
XM_011545690.2 | XP_011543992.1 | 7 | 1317 | |||
XM_011545693.2 | XP_011543995.1 | 7 | 1316 | |||
XM_017022814.1 | XP_016878303.1 | 7 | 1317 | |||
XM_017022815.1 | XP_016878304.1 | 7 | 837 | |||
XM_017022816.1 | XP_016878305.1 | 7 | 1317 | |||
XM_017022817.1 | XP_016878306.1 | 6 | 1229 | |||
XM_017022818.1 | XP_016878307.1 | 6 | 2554 | |||
XM_017022819.1 | XP_016878308.1 | 7 | 1244 | |||
XM_017022820.1 | XP_016878309.1 | 7 | 961 | |||
XM_017022821.1 | XP_016878310.1 | 6 | 1827 | |||
XM_017022822.1 | XP_016878311.1 | 7 | 2830 | |||
XM_017022823.1 | XP_016878312.1 | 5 | 2466 | |||
XM_017022824.1 | XP_016878313.1 | 7 | 1243 | |||
XM_017022825.1 | XP_016878314.1 | 7 | 961 | |||
XM_017022826.1 | XP_016878315.1 | 6 | 778 | |||
XM_017022827.1 | XP_016878316.1 | 2 | 525 | |||
XM_017022828.1 | XP_016878317.1 | 7 | 1316 | |||
GenBank mRNA | BC171825.1 | - | 6 | 1403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
