siRNA Details
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130464.2 | NP_569731.2 | 7 | 706 | ||
GenBank mRNA | AB209632.1 | - | 6 | 3273 | ||
AK128772.1 | - | 2 | 1826 | |||
AK294452.1 | - | 3 | 269 | |||
AK295170.1 | - | 7 | 705 | |||
AK300277.1 | - | 23 | ||||
AK303500.1 | - | 3 | 200 | |||
AK304162.1 | - | 5 | 488 | |||
AK308629.1 | - | 1 | 358 | |||
AK316459.1 | - | 3 | 135 | |||
BC036263.1 | - | 7 | 706 | 3903371 | ||
BC094882.1 | - | 6 | 633 | 3093162 | ||
BM557638.1 | - | 3 | 280 | |||
KF175176.1 | - | 6 | 630 | |||
KF175178.1 | - | 6 | 531 | |||
KF175179.1 | - | 6 | 573 | |||
KF175180.1 | - | 6 | 573 | |||
KF175181.1 | - | 6 | 717 | |||
KF175182.1 | - | 6 | 573 | |||
KF175184.1 | - | 5 | 456 | |||
KF175185.1 | - | 6 | 573 | |||
KF175186.1 | - | 5 | 447 | |||
KF175187.1 | - | 6 | 573 | |||
KF175188.1 | - | 6 | 534 | |||
KF175189.1 | - | 6 | 534 | |||
KF175190.1 | - | 6 | 573 | |||
KF175191.1 | - | 6 | 534 | |||
KF175192.1 | - | 6 | 534 | |||
KF175193.1 | - | 6 | 537 | |||
KF175194.1 | - | 6 | 537 | |||
KF175196.1 | - | 6 | 534 | |||
KF175197.1 | - | 6 | 534 | |||
KF175198.1 | - | 6 | 630 | |||
KF175199.1 | - | 6 | 630 | |||
KF175200.1 | - | 6 | 630 | |||
KF175201.1 | - | 6 | 573 | |||
KF175202.1 | - | 6 | 534 | |||
KF175203.1 | - | 6 | 630 | |||
KF175205.1 | - | 6 | 630 | |||
KF175206.1 | - | 6 | 630 | |||
KF175215.1 | - | 8 | 892 | |||
KF175216.1 | - | 7 | 907 | |||
KF175218.1 | - | 5 | 644 | |||
KF175223.1 | - | 6 | 630 | |||
KF175224.1 | - | 6 | 573 |
Gene Symbol: LOC613037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321892.1 | NP_001308821.1 | 8 | 823 | ||
XM_017023514.1 | XP_016879003.1 | 8 | 1112 | |||
XM_017023515.1 | XP_016879004.1 | 9 | 1131 | |||
XM_017023516.1 | XP_016879005.1 | 8 | 1158 | |||
XM_017023517.1 | XP_016879006.1 | 8 | 889 | |||
XM_017023518.1 | XP_016879007.1 | 8 | 977 | |||
XM_017023519.1 | XP_016879008.1 | 8 | 1034 | |||
XM_017023520.1 | XP_016879009.1 | 3 | 2098 | |||
XM_017023521.1 | XP_016879010.1 | 4 | 353 |
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KF175208.1 | - | 7 | 777 |
Gene Symbol: NPIPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310137.1 | NP_001297066.1 | 6 | 630 | ||
GenBank mRNA | KF175195.1 | - | 6 | 534 | ||
KF175204.1 | - | 6 | 573 |
Gene Symbol: NPIPB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310148.1 | NP_001297077.1 | 7 | 855 | ||
XM_017023244.1 | XP_016878733.1 | 6 | 2866 | |||
XM_017023245.1 | XP_016878734.1 | 7 | 2703 | |||
XM_017023246.1 | XP_016878735.1 | 7 | 2624 | |||
XM_017023247.1 | XP_016878736.1 | 6 | 857 |
Gene Symbol: NPIPB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135865.1 | NP_001129337.1 | 6 | 909 | ||
GenBank mRNA | AK296301.1 | - | 190 | |||
AK296338.1 | - | 6 | 750 | |||
AK303264.1 | - | 3 | 222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
