siRNA Details
Gene Symbol: POM121L1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024591.1 | 1 | 1371 | |||
GenBank mRNA | AK292412.1 | - | 1 | 1303 |
Gene Symbol: LOC101929599 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010105.1 | XP_016865594.1 | 1 | 84 |
Gene Symbol: LOC102724728 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136575.1 | 1 | 1299 | |||
GenBank mRNA | AK302597.1 | - | 1 | 1299 |
Gene Symbol: LOC107986351 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010104.1 | XP_016865593.1 | 1 | 69 |
Gene Symbol: LOC441081 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073404.1 | 1 | 2817 |
Gene Symbol: LOC728488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK296222.1 | - | 1 | 402 | ||
AK311627.1 | - | 1 | 678 |
Gene Symbol: POM121L10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024593.1 | 1 | 1299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
