siRNA Details
Gene Symbol: CSAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129826.2 | NP_001123298.1 | 1 | 224 | ||
NM_001129828.2 | NP_001123300.1 | 1 | 224 | |||
GenBank mRNA | AF080246.1 | - | 1 | 224 | ||
AF136715.1 | - | 1 | 224 | |||
BC119734.1 | - | 1 | 197 | 40119148 | ||
BC119735.1 | - | 1 | 196 | 40119150 |
Gene Symbol: CSAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724857.3 | XP_006724920.1 | 1 | 350 | ||
XM_017030023.1 | XP_016885512.1 | 1 | 350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
