siRNA Details
Gene Symbol: ATP7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000052.6 | NP_000043.4 | 11 | 2652 | ||
NM_001282224.1 | NP_001269153.1 | 10 | 2418 | |||
GenBank mRNA | AB117973.1 | - | 11 | 2579 | ||
L06133.1 | - | 11 | 2596 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
