siRNA Details
Gene Symbol: GH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002059.4 | NP_002050.1 | 2 | 169 | ||
NM_022556.3 | NP_072050.1 | 2 | 169 | |||
NM_022557.3 | NP_072051.1 | 2 | 169 | |||
NM_022558.3 | NP_072052.1 | 2 | 169 | |||
GenBank mRNA | AB451332.1 | - | 1 | 28 | ||
AB451477.1 | - | 1 | 28 | |||
AF006060.1 | - | 2 | 40 | |||
AF006061.1 | - | 1 | 28 | |||
AK312194.1 | - | 2 | 91 | |||
AU139016.1 | - | 2 | 169 | |||
BC020760.1 | - | 2 | 90 | 4720475 | ||
CR541902.1 | - | 1 | 28 | |||
CR541924.1 | - | 1 | 28 | |||
J03756.1 | - | 2 | 89 | |||
M38451.1 | - | 2 | 40 |
Gene Symbol: CSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317.5 | NP_001308.1 | 2 | 178 | ||
GenBank mRNA | AK291587.1 | - | 2 | 97 | ||
AU134786.2 | - | 2 | 178 | |||
BC002717.2 | - | 2 | 71 | 3631916 | ||
BC005921.1 | - | 2 | 90 | 4043768 | ||
BC020756.1 | - | 2 | 146 | 4769769 | ||
BC057768.1 | - | 2 | 90 | 30337725 | ||
BC062775.1 | - | 2 | 94 | 4604848 | ||
BG436049.1 | - | 2 | 91 | |||
BT006926.1 | - | 1 | 28 | |||
J00118.1 | - | 2 | 57 |
Gene Symbol: CSH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020991.3 | NP_066271.1 | 2 | 143 | ||
NM_022644.3 | NP_072170.1 | 2 | 143 | |||
NM_022645.2 | NP_072171.1 | 2 | 143 | |||
GenBank mRNA | BC022044.1 | - | 2 | 94 | 4769046 | |
BC035965.1 | - | 2 | 90 | 4731880 | ||
BC119748.2 | - | 2 | 87 | 40119242 | ||
V00573.1 | - | 2 | 52 |
Gene Symbol: CSHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321068.1 | NP_001307997.1 | 2 | 160 | ||
NM_001321069.1 | NP_001307998.1 | 2 | 160 | |||
NM_022579.2 | NP_072101.1 | 2 | 160 | |||
NM_022581.2 | NP_072103.1 | 2 | 160 | |||
XM_011524344.1 | XP_011522646.1 | 2 | 90 | |||
GenBank mRNA | AK300190.1 | - | 2 | 90 | ||
AX772828.1 | - | 2 | 174 | |||
BX426214.2 | - | 2 | 73 | |||
DR004701.1 | - | 2 | 70 |
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000515.4 | NP_000506.2 | 2 | 104 | ||
NM_022559.3 | NP_072053.1 | 2 | 104 | |||
NM_022560.3 | NP_072054.1 | 2 | 104 | |||
GenBank mRNA | A00469.1 | - | 2 | 67 | ||
AF110644.1 | - | 2 | 38 | |||
AF185611.1 | - | 2 | 46 | |||
AV749353.1 | - | 2 | 104 | |||
AY613431.1 | - | 2 | 90 | |||
AY613432.1 | - | 2 | 90 | |||
BC062475.1 | - | 2 | 69 | 30393581 | ||
BC075012.2 | - | 2 | 55 | 30915404 | ||
BC075013.2 | - | 2 | 55 | 30915617 | ||
BC090045.1 | - | 2 | 71 | 30394115 | ||
CD108592.1 | - | 2 | 70 | |||
M14398.1 | - | 1 | 28 | |||
V00519.1 | - | 2 | 68 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
