siRNA Details
Gene Symbol: SPDYE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004351.4 | NP_001004351.3 | 2 | 393 | ||
XM_011516234.2 | XP_011514536.1 | 2 | 1296 | |||
XM_011516235.2 | XP_011514537.1 | 2 | 1296 | |||
GenBank mRNA | BC056606.1 | - | 2 | 394 | 4838469 | |
BC100972.2 | - | 1 | 194 |
Gene Symbol: LOC105371844 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011525567.2 | XP_011523869.1 | 2 | 693 |
Gene Symbol: SPDYE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175064.2 | NP_778234.2 | 1 | 279 | ||
XM_005249719.3 | XP_005249776.1 | 3 | 790 | |||
XM_017012015.1 | XP_016867504.1 | 3 | 728 | |||
XM_017012016.1 | XP_016867505.1 | 2 | 140 | |||
GenBank mRNA | AF412027.1 | - | 1 | 288 | ||
AL137266.1 | - | 1 | 61 |
Gene Symbol: SPDYE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716210.3 | XP_006716273.1 | 2 | 763 | ||
XM_017012933.1 | XP_016868422.1 | 2 | 767 |
Gene Symbol: SPDYE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716211.3 | XP_006716274.1 | 1 | 417 | ||
XM_017012887.1 | XP_016868376.1 | 1 | 417 |
Gene Symbol: SPDYE17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716212.3 | XP_006716275.1 | 2 | 767 | ||
XM_017012888.1 | XP_016868377.1 | 2 | 767 |
Gene Symbol: SPDYE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250742.4 | XP_005250799.1 | 2 | 288 | ||
XM_005250743.4 | XP_005250800.1 | 1 | 143 | |||
XM_011516756.2 | XP_011515058.1 | 2 | 288 |
Gene Symbol: SPDYE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031618.3 | NP_001026789.2 | 3 | 737 | ||
XM_011516237.2 | XP_011514539.1 | 3 | 736 | |||
XM_017012223.1 | XP_016867712.1 | 3 | 736 | |||
GenBank mRNA | AK302217.1 | - | 3 | 737 | ||
JQ023160.1 | - | 2 | 263 |
Gene Symbol: SPDYE2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166339.1 | NP_001159811.1 | 3 | 737 | ||
XM_005250093.3 | XP_005250150.1 | 2 | 1305 | |||
XM_011515702.2 | XP_011514004.1 | 2 | 1305 |
Gene Symbol: SPDYE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146210.2 | NP_001139682.2 | 2 | 684 |
Gene Symbol: SPDYE8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003664.2 | 3 | 804 | |||
GenBank mRNA | BC045636.1 | - | 3 | 807 | ||
BC068610.1 | - | 3 | 807 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
