siRNA Details
Gene Symbol: TSPY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022573.2 | NP_072095.2 | 3 | 636 | ||
XM_005262569.4 | XP_005262626.1 | 3 | 596 | |||
XM_017030062.1 | XP_016885551.1 | 3 | 596 | |||
GenBank mRNA | AK307425.1 | - | 3 | 541 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
