siRNA Details
Gene Symbol: MTURN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152793.2 | NP_690006.2 | 2 | 445 | ||
XM_005249652.3 | XP_005249709.1 | 2 | 388 | |||
GenBank mRNA | AK294982.1 | - | 2 | 175 | ||
AL834391.1 | - | 2 | 379 | |||
AY054121.1 | - | 2 | 241 | |||
BC062423.1 | - | 2 | 196 | |||
BI560774.1 | - | 2 | 449 | |||
DT217816.1 | - | 2 | 380 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
