siRNA Details
Gene Symbol: OR1J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123016.1 | - | 4 | 496 |
Gene Symbol: MGC72080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002822.3 | 4 | 440 | |||
GenBank mRNA | BC029615.1 | - | 4 | 450 | ||
BC042035.1 | - | 3 | 379 |
Gene Symbol: OR10G4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004462.1 | NP_001004462.1 | 1 | 164 |
Gene Symbol: OR10G7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004463.1 | NP_001004463.1 | 1 | 164 |
Gene Symbol: OR10G9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001953.1 | NP_001001953.1 | 1 | 164 |
Gene Symbol: OR1M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004456.1 | NP_001004456.1 | 1 | 170 | ||
GenBank mRNA | BC137551.1 | - | 1 | 236 | 9021551 | |
BC137555.1 | - | 1 | 236 | 9021555 |
Gene Symbol: OR1N1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012363.1 | NP_036495.1 | 1 | 164 |
Gene Symbol: OR2T4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004696.1 | NP_001004696.1 | 1 | 266 |
Gene Symbol: OR2W3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001957.2 | NP_001001957.2 | 1 | 170 | ||
GenBank mRNA | BC146848.1 | - | 1 | 201 | 9056776 |
Gene Symbol: OR7A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005190.1 | NP_001005190.1 | 1 | 170 |
Gene Symbol: OR7A17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030901.1 | NP_112163.1 | 1 | 170 | ||
GenBank mRNA | BC101587.1 | - | 1 | 267 | 8069093 | |
BC117357.1 | - | 1 | 267 | 40125908 |
Gene Symbol: OR7A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017506.1 | NP_059976.1 | 1 | 393 | ||
XM_011527912.2 | XP_011526214.1 | 2 | 418 | |||
XM_011527913.2 | XP_011526215.1 | 2 | 344 | |||
XM_017026583.1 | XP_016882072.1 | 2 | 307 | |||
GenBank mRNA | AK312475.1 | - | 2 | 259 | ||
BC104809.1 | - | 1 | 228 | 8143812 |
Gene Symbol: OR7C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198944.1 | NP_945182.1 | 1 | 170 | ||
XM_005259853.3 | XP_005259910.1 | 3 | 674 | |||
XM_011527915.2 | XP_011526217.1 | 5 | 1095 | |||
GenBank mRNA | BC117212.1 | - | 1 | 245 | 40125763 |
Gene Symbol: OR7C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012377.1 | NP_036509.1 | 1 | 170 |
Gene Symbol: OR7D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175883.2 | NP_787079.1 | 1 | 358 | ||
GenBank mRNA | AK095468.1 | - | 1 | 358 |
Gene Symbol: OR7D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005191.2 | NP_001005191.1 | 1 | 204 | ||
GenBank mRNA | BC137147.1 | - | 1 | 204 | 9021144 | |
BC137162.1 | - | 1 | 204 | 9021159 |
Gene Symbol: OR7E12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_044999.1 | 1 | 342 |
Gene Symbol: OR7E14P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045002.1 | 1 | 339 | |||
GenBank mRNA | BC008657.2 | - | 1 | 194 |
Gene Symbol: OR7E24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079935.1 | NP_001073404.1 | 1 | 224 | ||
GenBank mRNA | BC146920.1 | - | 1 | 338 | 9056848 |
Gene Symbol: OR7G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005192.2 | NP_001005192.2 | 1 | 170 |
Gene Symbol: OR7G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005193.1 | NP_001005193.1 | 1 | 233 |
Gene Symbol: OR7G3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001958.1 | NP_001001958.1 | 1 | 170 |
Gene Symbol: TRIM58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX537432.1 | - | 3 | 427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
