siRNA Details
Gene Symbol: IGKV3-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ006111.1 | - | 248 | |||
AJ006112.1 | - | 248 | ||||
AJ006114.1 | - | 248 | ||||
AJ006115.1 | - | 248 | ||||
AJ006117.1 | - | 248 | ||||
AJ006118.1 | - | 248 | ||||
AJ006119.1 | - | 248 | ||||
AJ006120.1 | - | 248 |
Gene Symbol: IGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX640854.1 | - | 2 | 385 |
Gene Symbol: IGHG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127409.1 | - | 2 | 391 | ||
AK128301.1 | - | 2 | 342 | |||
AK128313.1 | - | 2 | 384 | |||
BC018747.1 | - | 2 | 367 | |||
BC090938.1 | - | 2 | 365 |
Gene Symbol: IGHM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC015760.1 | - | 2 | 364 | ||
BC020240.1 | - | 2 | 375 | |||
BC041037.1 | - | 2 | 353 | |||
BC073758.1 | - | 2 | 374 |
Gene Symbol: IGHV3-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF231396.1 | - | 1 | 248 | ||
AJ519334.1 | - | 1 | 278 | |||
X14584.1 | - | 2 | 329 |
Gene Symbol: IGHV3-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF035024.1 | - | 1 | 248 |
Gene Symbol: IGHV3-48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF035023.1 | - | 2 | 248 |
Gene Symbol: IGHV3-69-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF460663.1 | - | 101 | |||
AF460692.1 | - | 1 | 101 | |||
AF460695.1 | - | 101 | ||||
AF460697.1 | - | 101 | ||||
AF460706.1 | - | 1 | 101 | |||
AF460787.1 | - | 101 | ||||
AF460807.1 | - | 101 | ||||
GU552754.1 | - | 158 | ||||
JX420391.1 | - | 1 | 143 | |||
JX420392.1 | - | 1 | 143 |
Gene Symbol: IGHV3-71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KP092035.1 | - | 1 | 183 |
Gene Symbol: IGHV3OR16-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF209733.1 | - | 1 | 193 | ||
AY003842.1 | - | 1 | 185 | |||
DQ454529.1 | - | 1 | 161 | |||
EU099108.1 | - | 1 | 200 |
Gene Symbol: IGHV4-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY172959.1 | - | 2 | 305 |
Gene Symbol: SCFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | Y13056.1 | - | 248 |
Gene Symbol: ZCWPW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB087877.1 | - | 1 | 262 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
