siRNA Details
Gene Symbol: ZFP64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319146.1 | NP_001306075.1 | 5 | 843 | ||
NM_018197.2 | NP_060667.2 | 6 | 1137 | |||
NM_022088.4 | NP_071371.3 | 5 | 975 | |||
NM_199426.1 | NP_955458.1 | 6 | 1131 | |||
NM_199427.2 | NP_955459.2 | 9 | 1602 | |||
XM_005260449.1 | XP_005260506.1 | 5 | 554 | |||
XM_006723822.1 | XP_006723885.1 | 5 | 772 | |||
XM_011528900.1 | XP_011527202.1 | 7 | 1028 | |||
XM_017027945.1 | XP_016883434.1 | 9 | 1388 | |||
XM_017027946.1 | XP_016883435.1 | 5 | 697 | |||
XM_017027947.1 | XP_016883436.1 | 8 | 1244 | |||
XM_017027948.1 | XP_016883437.1 | 6 | 676 | |||
XM_017027949.1 | XP_016883438.1 | 7 | 995 | |||
GenBank mRNA | AK001596.1 | - | 6 | 927 | ||
AK001744.1 | - | 5 | 702 | |||
AK022690.1 | - | 5 | 843 | |||
AK090547.1 | - | 7 | 799 | |||
BC012759.2 | - | 6 | 1137 | 3634346 | ||
BC021087.1 | - | 9 | 1323 | |||
BC041622.1 | - | 6 | 1137 | 3633938 | ||
BT009760.1 | - | 9 | 1253 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
