siRNA Details
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282524.1 | NP_001269453.1 | 4 | 462 | ||
XM_005255741.3 | XP_005255798.1 | 5 | 1274 | |||
XM_005255742.4 | XP_005255799.1 | 5 | 1020 | |||
XM_011545929.2 | XP_011544231.1 | 6 | 1094 | |||
XM_011545931.2 | XP_011544233.1 | 5 | 727 | |||
XM_017023628.1 | XP_016879117.1 | 5 | 477 | |||
XM_017023629.1 | XP_016879118.1 | 5 | 1020 | |||
XM_017023630.1 | XP_016879119.1 | 6 | 747 | |||
XM_017023631.1 | XP_016879120.1 | 4 | 1091 | |||
XM_017023632.1 | XP_016879121.1 | 5 | 477 | |||
GenBank mRNA | BC144439.1 | - | 4 | 423 | ||
BC157836.1 | - | 4 | 1112 | 9057055 | ||
KF175222.1 | - | 4 | 424 |
Gene Symbol: LOC613037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321892.1 | NP_001308821.1 | 6 | 563 | ||
XM_017023514.1 | XP_016879003.1 | 6 | 852 | |||
XM_017023515.1 | XP_016879004.1 | 7 | 871 | |||
XM_017023516.1 | XP_016879005.1 | 6 | 898 | |||
XM_017023517.1 | XP_016879006.1 | 6 | 629 | |||
XM_017023518.1 | XP_016879007.1 | 6 | 717 | |||
XM_017023519.1 | XP_016879008.1 | 6 | 774 | |||
XM_017023521.1 | XP_016879010.1 | 1 | 24 | |||
GenBank mRNA | BQ433543.1 | - | 486 | |||
DB231720.1 | - | 5 | 432 |
Gene Symbol: NPIPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310137.1 | NP_001297066.1 | 4 | 370 | ||
GenBank mRNA | KF175195.1 | - | 4 | 274 | ||
KF175204.1 | - | 4 | 313 |
Gene Symbol: NPIPB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306094.1 | NP_001293023.1 | 4 | 392 | ||
XM_011523496.2 | XP_011521798.1 | 5 | 448 | |||
XM_011523498.2 | XP_011521800.1 | 5 | 482 | |||
XM_017023248.1 | XP_016878737.1 | 5 | 885 | |||
XM_017023249.1 | XP_016878738.1 | 4 | 702 |
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130464.2 | NP_569731.2 | 5 | 446 | ||
GenBank mRNA | AB209632.1 | - | 4 | 3013 | ||
AK295170.1 | - | 5 | 445 | |||
AK304162.1 | - | 3 | 228 | |||
BC036263.1 | - | 5 | 446 | 3903371 | ||
BC094882.1 | - | 4 | 373 | 3093162 | ||
BM557638.1 | - | 1 | 20 | |||
DC342292.1 | - | 4 | 550 | |||
KF175175.1 | - | 4 | 313 | |||
KF175176.1 | - | 4 | 370 | |||
KF175177.1 | - | 4 | 370 | |||
KF175178.1 | - | 4 | 271 | |||
KF175179.1 | - | 4 | 313 | |||
KF175180.1 | - | 4 | 313 | |||
KF175181.1 | - | 4 | 457 | |||
KF175182.1 | - | 4 | 313 | |||
KF175183.1 | - | 3 | 205 | |||
KF175184.1 | - | 3 | 196 | |||
KF175185.1 | - | 4 | 313 | |||
KF175186.1 | - | 3 | 187 | |||
KF175187.1 | - | 4 | 313 | |||
KF175188.1 | - | 4 | 274 | |||
KF175189.1 | - | 4 | 274 | |||
KF175190.1 | - | 4 | 313 | |||
KF175191.1 | - | 4 | 274 | |||
KF175192.1 | - | 4 | 274 | |||
KF175193.1 | - | 4 | 277 | |||
KF175194.1 | - | 4 | 277 | |||
KF175196.1 | - | 4 | 274 | |||
KF175197.1 | - | 4 | 274 | |||
KF175198.1 | - | 4 | 370 | |||
KF175199.1 | - | 4 | 370 | |||
KF175200.1 | - | 4 | 370 | |||
KF175201.1 | - | 4 | 313 | |||
KF175202.1 | - | 4 | 274 | |||
KF175203.1 | - | 4 | 370 | |||
KF175205.1 | - | 4 | 370 | |||
KF175206.1 | - | 4 | 370 | |||
KF175215.1 | - | 6 | 632 | |||
KF175216.1 | - | 5 | 647 | |||
KF175218.1 | - | 4 | 275 | |||
KF175223.1 | - | 4 | 370 | |||
KF175224.1 | - | 4 | 313 |
Gene Symbol: NPIPB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310148.1 | NP_001297077.1 | 5 | 595 | ||
XM_017023244.1 | XP_016878733.1 | 4 | 2606 | |||
XM_017023245.1 | XP_016878734.1 | 5 | 2443 | |||
XM_017023246.1 | XP_016878735.1 | 5 | 2364 | |||
XM_017023247.1 | XP_016878736.1 | 4 | 597 |
Gene Symbol: NPIPB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135865.1 | NP_001129337.1 | 4 | 649 | ||
GenBank mRNA | AK296338.1 | - | 4 | 490 |
Gene Symbol: NPIPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310136.1 | NP_001297065.1 | 4 | 424 | ||
XM_017023622.1 | XP_016879111.1 | 5 | 548 | |||
XM_017023623.1 | XP_016879112.1 | 5 | 548 | |||
XM_017023624.1 | XP_016879113.1 | 5 | 548 | |||
XM_017023625.1 | XP_016879114.1 | 5 | 528 | |||
XM_017023626.1 | XP_016879115.1 | 6 | 1167 | |||
XM_017023627.1 | XP_016879116.1 | 5 | 548 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287250.1 | NP_001274179.1 | 4 | 462 | ||
NM_001287251.1 | NP_001274180.1 | 5 | 548 | |||
XM_003118719.7 | XP_003118767.2 | 5 | 1023 | |||
XM_011545690.2 | XP_011543992.1 | 6 | 1112 | |||
XM_011545693.2 | XP_011543995.1 | 6 | 1111 | |||
XM_017022814.1 | XP_016878303.1 | 6 | 1112 | |||
XM_017022815.1 | XP_016878304.1 | 6 | 632 | |||
XM_017022816.1 | XP_016878305.1 | 6 | 1112 | |||
XM_017022817.1 | XP_016878306.1 | 5 | 1024 | |||
XM_017022818.1 | XP_016878307.1 | 5 | 2349 | |||
XM_017022819.1 | XP_016878308.1 | 6 | 1039 | |||
XM_017022820.1 | XP_016878309.1 | 6 | 756 | |||
XM_017022821.1 | XP_016878310.1 | 5 | 1622 | |||
XM_017022822.1 | XP_016878311.1 | 6 | 2625 | |||
XM_017022823.1 | XP_016878312.1 | 4 | 2261 | |||
XM_017022824.1 | XP_016878313.1 | 6 | 1038 | |||
XM_017022825.1 | XP_016878314.1 | 6 | 756 | |||
XM_017022826.1 | XP_016878315.1 | 5 | 573 | |||
XM_017022828.1 | XP_016878317.1 | 6 | 1111 | |||
GenBank mRNA | BC171825.1 | - | 5 | 1198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
