siRNA Details
Gene Symbol: DNAH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207437.3 | NP_997320.2 | 28, 29 | 4803 | ||
XM_005253555.3 | XP_005253612.1 | 29, 30 | 5466 | |||
XM_011538014.2 | XP_011536316.1 | 29, 30 | 5466 | |||
XM_011538015.2 | XP_011536317.1 | 29, 30 | 5466 | |||
XM_011538016.2 | XP_011536318.1 | 29, 30 | 5466 | |||
XM_011538017.2 | XP_011536319.1 | 29, 30 | 5466 | |||
XM_011538019.2 | XP_011536321.1 | 11, 12 | 2065 | |||
XM_011538020.2 | XP_011536322.1 | 29, 30 | 5474 | |||
XM_017018960.1 | XP_016874449.1 | 29, 30 | 5466 | |||
XM_017018961.1 | XP_016874450.1 | 28, 29 | 5157 | |||
XM_017018962.1 | XP_016874451.1 | 11, 12 | 1987 | |||
GenBank mRNA | AB095937.2 | - | 3, 4 | 518 | ||
AB290168.1 | - | 8, 9 | 1322 | |||
AK125796.1 | - | 2, 3 | 320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
