siRNA Details
Gene Symbol: PRKCQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242413.2 | NP_001229342.1 | 17 | 2967 | ||
NM_001282644.1 | NP_001269573.1 | 18 | 3154 | |||
NM_001282645.1 | NP_001269574.1 | 17 | 3095 | |||
NM_001323265.1 | NP_001310194.1 | 18 | 3211 | |||
NM_001323266.1 | NP_001310195.1 | 17 | 3093 | |||
NM_006257.4 | NP_006248.1 | 18 | 3156 | |||
GenBank mRNA | BX647657.1 | - | 17 | 3241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
