siRNA Details
Gene Symbol: Jak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190830.1 | NP_001177759.1 | 14 | 2025 | ||
NM_010589.6 | NP_034719.2 | 14 | 2025 | |||
GenBank mRNA | AK038268.1 | - | 13 | 1836 | ||
AK088365.1 | - | 14 | 1874 | |||
AK142178.1 | - | 14 | 1863 | |||
AK156646.1 | - | 14 | 1847 | |||
AK164520.1 | - | 14 | 1865 | |||
BC105577.1 | - | 13 | 1828 | 3968386 | ||
BC131646.1 | - | 13 | 1836 | 40110146 | ||
BC131647.1 | - | 13 | 1836 | 40110148 | ||
L32955.1 | - | 13 | 1824 | |||
L33768.1 | - | 11 | 2466 | |||
L40172.1 | - | 14 | 1886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
