siRNA Details
Gene Symbol: Jak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190830.1 | NP_001177759.1 | 12 | 1784 | ||
NM_010589.6 | NP_034719.2 | 12 | 1784 | |||
GenBank mRNA | AK038268.1 | - | 11 | 1595 | ||
AK088365.1 | - | 12 | 1633 | |||
AK142178.1 | - | 12 | 1622 | |||
AK156646.1 | - | 12 | 1606 | |||
AK164520.1 | - | 12 | 1624 | |||
BC105577.1 | - | 11 | 1587 | 3968386 | ||
BC131646.1 | - | 11 | 1595 | 40110146 | ||
BC131647.1 | - | 11 | 1595 | 40110148 | ||
L32955.1 | - | 11 | 1583 | |||
L33768.1 | - | 9 | 2225 | |||
L40172.1 | - | 12 | 1645 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
