siRNA Details
Gene Symbol: IL12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000882.3 | NP_000873.2 | 5, 6 | 678 | ||
GenBank mRNA | AF101062.1 | - | 5, 6 | 457 | ||
AF180562.1 | - | 5, 6 | 516 | |||
BC104982.1 | - | 5, 6 | 610 | 8143985 | ||
BC104984.1 | - | 5, 6 | 610 | 8143987 | ||
M65271.1 | - | 5, 6 | 524 | |||
M65291.1 | - | 5, 6 | 558 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
