siRNA Details
Gene Symbol: Ripk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138952.3 | NP_620402.1 | 5, 6 | 791 | ||
GenBank mRNA | AF461040.1 | - | 5, 6 | 689 | ||
AF487539.1 | - | 5, 6 | 689 | |||
AK159593.1 | - | 5, 6 | 815 | |||
AK172319.1 | - | 5, 6 | 813 | |||
AK172417.1 | - | 5, 6 | 813 | |||
BC025611.1 | - | 3, 4 | 309 | |||
BC050845.1 | - | 5, 6 | 804 | |||
BC058088.1 | - | 5, 6 | 936 | |||
BC069878.1 | - | 5, 6 | 816 | 5316066 |

More Information
Other Species Targeted

Gene Ontology Categories:
