siRNA Details
Gene Symbol: Ripk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138952.3 | NP_620402.1 | 7 | 984 | ||
GenBank mRNA | AF461040.1 | - | 7 | 882 | ||
AF487539.1 | - | 7 | 882 | |||
AK159593.1 | - | 7 | 1008 | |||
AK172319.1 | - | 7 | 1006 | |||
AK172417.1 | - | 7 | 1006 | |||
BC025611.1 | - | 5 | 502 | |||
BC050845.1 | - | 7 | 998 | |||
BC058088.1 | - | 7 | 1129 | |||
BC069878.1 | - | 7 | 1009 | 5316066 |

More Information
Other Species Targeted

Gene Ontology Categories:
