siRNA Details
Gene Symbol: HINT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005340.6 | NP_005331.1 | 1 | 17 | ||
NR_024610.2 | 1 | 17 | ||||
NR_024611.2 | 1 | 17 | ||||
NR_073488.1 | 1 | 17 | ||||
NR_134494.1 | 1 | 17 | ||||
NR_134495.1 | 1 | 17 | ||||
GenBank mRNA | BF967059.1 | - | 1 | 158 | ||
BG708765.1 | - | 1 | 22 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
