siRNA Details
Gene Symbol: NCAPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281710.1 | NP_001268639.1 | 1 | 57 | ||
NM_001281711.1 | NP_001268640.1 | 1 | 57 | |||
NM_001281712.1 | NP_001268641.1 | 1 | 57 | |||
NM_015341.4 | NP_056156.2 | 1 | 57 | |||
GenBank mRNA | BC024211.2 | - | 11 | 3504782 | ||
BI559873.1 | - | 1 | 45 | |||
BI560881.1 | - | 1 | 62 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
