siRNA Details
Gene Symbol: NCAPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281710.1 | NP_001268639.1 | 18 | 2545 | ||
NM_001281711.1 | NP_001268640.1 | 18 | 2506 | |||
NM_001281712.1 | NP_001268641.1 | 17 | 2325 | |||
NM_015341.4 | NP_056156.2 | 18 | 2578 | |||
XM_005263908.3 | XP_005263965.1 | 18 | 2531 | |||
XM_006712388.3 | XP_006712451.1 | 18 | 2425 | |||
GenBank mRNA | AK303725.1 | - | 17 | 2270 | ||
BC024211.2 | - | 18 | 2532 | 3504782 | ||
D38553.1 | - | 18 | 2508 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
