siRNA Details
Gene Symbol: Tpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057208.2 | NP_476556.2 | 8 | 1019 | ||
XM_006232576.2 | XP_006232638.1 | 9 | 1215 | |||
XM_006232577.2 | XP_006232639.1 | 9 | 1215 | |||
XM_006232584.2 | XP_006232646.1 | 8 | 1121 | |||
GenBank mRNA | L24777.1 | - | 8 | 1019 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
