siRNA Details
Gene Symbol: DCHS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017639.3 | NP_060109.2 | 23, 24 | 6002 | ||
XM_011532045.1 | XP_011530347.1 | 18, 19 | 7790 | |||
XM_011532046.2 | XP_011530348.1 | 17, 18 | 5446 | |||
XM_011532049.2 | XP_011530351.1 | 10, 11 | 3296 | |||
GenBank mRNA | AK000054.1 | - | 1, 2 | 299 | ||
AK095103.1 | - | 6, 7 | 1339 | |||
AK225200.1 | - | 1, 2 | 299 | |||
AY354497.1 | - | 23, 24 | 6002 | |||
BX647835.1 | - | 6, 7 | 978 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
