siRNA Details
Gene Symbol: Olr1096 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000485.1 | NP_001000485.1 | 1 | 203 | ||
XM_008762647.1 | XP_008760869.1 | 1 | 739 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1096 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000485.1 | NP_001000485.1 | 1 | 203 | ||
XM_008762647.1 | XP_008760869.1 | 1 | 739 |