siRNA Details
Gene Symbol: Ablim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001514.2 | NP_001001514.1 | 6 | 769 | ||
NM_001177695.1 | NP_001171166.1 | 6 | 769 | |||
XM_006251135.2 | XP_006251197.1 | 6 | 1326 | |||
XM_006251137.2 | XP_006251199.1 | 6 | 1325 | |||
XM_006251138.2 | XP_006251200.1 | 6 | 1325 | |||
XM_006251139.2 | XP_006251201.1 | 6 | 1325 | |||
XM_008770327.1 | XP_008768549.1 | 6 | 766 | |||
XM_008770328.1 | XP_008768550.1 | 6 | 1325 | |||
XM_008770329.1 | XP_008768551.1 | 6 | 1324 | |||
XM_008770330.1 | XP_008768552.1 | 6 | 1325 | |||
GenBank mRNA | AJ703892.2 | - | 6 | 769 | ||
AJ703893.2 | - | 6 | 769 |
