siRNA Details
Gene Symbol: GPATCH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002909.3 | NP_001002909.1 | 8 | 4988 | ||
NM_001304939.1 | NP_001291868.1 | 7 | 4913 | |||
NM_001304940.1 | NP_001291869.1 | 10 | 5129 | |||
NM_001304941.1 | NP_001291870.1 | 9 | 5006 | |||
NM_001304942.1 | NP_001291871.1 | 7 | 4915 | |||
NM_001304943.1 | NP_001291872.1 | 5 | 4869 | |||
XM_011524557.1 | XP_011522859.1 | 10 | 5062 | |||
XM_011524558.1 | XP_011522860.1 | 11 | 4881 | |||
XM_011524559.2 | XP_011522861.1 | 12 | 4959 | |||
XM_011524561.2 | XP_011522863.1 | 8 | 4720 | |||
XM_017024372.1 | XP_016879861.1 | 13 | 5004 | |||
XM_017024373.1 | XP_016879862.1 | 10 | 5839 | |||
XM_017024374.1 | XP_016879863.1 | 12 | 4922 | |||
XM_017024375.1 | XP_016879864.1 | 9 | 5017 | |||
XM_017024376.1 | XP_016879865.1 | 10 | 4877 | |||
XM_017024377.1 | XP_016879866.1 | 9 | 4854 | |||
XM_017024378.1 | XP_016879867.1 | 10 | 5035 | |||
XM_017024379.1 | XP_016879868.1 | 11 | 4917 | |||
XM_017024380.1 | XP_016879869.1 | 10 | 4855 | |||
GenBank mRNA | AB011125.2 | - | 7 | 6078 | ||
BC024147.2 | - | 1 | 1767 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001159492, BC035255, AK122307, AK142570.
Gene Ontology Categories:
