siRNA Details
Gene Symbol: C10orf90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004298.2 | NP_001004298.2 | 1, 2 | 151 | ||
GenBank mRNA | AK057500.1 | - | 1, 2 | 151 | ||
AL705062.1 | - | 1, 2 | 176 | |||
BC140898.1 | - | 1, 2 | 138 | 9021766 | ||
BC146869.1 | - | 1, 2 | 138 | 9056797 |
