siRNA Details
Gene Symbol: ZNF720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130913.1 | NP_001124385.1 | 5 | 2296 | ||
GenBank mRNA | BC018134.1 | - | 5 | 2251 | 3912539 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130913.1 | NP_001124385.1 | 5 | 2296 | ||
GenBank mRNA | BC018134.1 | - | 5 | 2251 | 3912539 |